Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380353.1 | NP_001368441.1 | 4 - 5 | 437 | 62 | |
NM_171913.6 | NP_741458.2 | 9 - 10 | 1046 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |