Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029536.5 | NP_001024707.1 | 7 - 8 | 2630 | 78 | |
GenBank mRNA | AF200706.1 | - | 7 - 8 | 2637 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |