Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064725.7 | NP_497126.1 | 5 - 6 | 1212 | 70 | |
NM_181901.2 | NP_871630.1 | 4 - 5 | 1150 | 70 | ||
GenBank mRNA | AY204179.1 | - | 5 - 6 | 1140 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |