Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373792.1 | NP_001359656.1 | 2 - 3 | 359 | 64 | |
NM_001381607.1 | NP_001367045.1 | 2 - 3 | 365 | 64 | ||
NM_064041.5 | NP_496442.1 | 2 - 3 | 359 | 64 | ||
NR_138514.1 | 2 - 3 | 977 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |