Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vhp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356833.4 | NP_001343753.1 | 5 - 6 | 878 | 88 | |
NM_001356834.3 | NP_001343752.1 | 4 - 5 | 653 | 88 | ||
NM_001356836.3 | NP_001343755.1 | 1 - 2 | 239 | 88 | ||
NM_001377798.2 | NP_001364748.1 | 5 - 6 | 869 | 88 | ||
NM_001377799.2 | NP_001364848.1 | 2 - 3 | 401 | 88 | ||
NM_062597.4 | NP_494998.1 | 3 - 4 | 923 | 88 | ||
NR_151559.1 | 2 - 3 | 401 | 88 | |||
GenBank mRNA | AY585194.1 | - | 5 - 6 | 866 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |