Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267523.2 | NP_001254452.1 | 5 - 6 | 1085 | 70 | |
NM_001267525.3 | NP_001254454.1 | 4 - 5 | 763 | 70 | ||
NM_001306640.4 | NP_001293569.1 | 6 - 7 | 1080 | 70 | ||
NM_001306641.2 | NP_001293570.1 | 5 - 6 | 1085 | 70 | ||
NM_001306642.3 | NP_001293571.1 | 8 - 9 | 1321 | 70 | ||
NM_001306643.3 | NP_001293572.1 | 7 - 8 | 1162 | 70 | ||
NM_001381668.1 | NP_001367248.1 | 8 - 9 | 1321 | 70 | ||
NM_001381670.1 | NP_001368261.1 | 6 - 7 | 1021 | 70 | ||
NM_001383996.1 | 8 - 9 | 1385 | 70 | |||
NM_064683.6 | NP_497084.2 | 3 - 4 | 871 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |