Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267523.2 | NP_001254452.1 | 8 - 9 | 1696 | 90 | |
NM_001267525.3 | NP_001254454.1 | 7 - 8 | 1374 | 90 | ||
NM_001306640.4 | NP_001293569.1 | 9 - 10 | 1691 | 90 | ||
NM_001306641.2 | NP_001293570.1 | 8 - 9 | 1696 | 90 | ||
NM_001306642.3 | NP_001293571.1 | 11 - 12 | 1932 | 90 | ||
NM_001306643.3 | NP_001293572.1 | 10 - 11 | 1773 | 90 | ||
NM_001381668.1 | NP_001367248.1 | 11 - 12 | 1932 | 90 | ||
NM_001381670.1 | NP_001368261.1 | 9 - 10 | 1632 | 90 | ||
NM_001383996.1 | 11 - 12 | 1996 | 90 | |||
NM_064683.6 | NP_497084.2 | 6 - 7 | 1482 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |