Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330963.2 | NP_001317775.1 | 6 - 7 | 1506 | 93 | |
NM_001330964.4 | NP_001317776.1 | 6 - 7 | 1512 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |