Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lntl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380241.1 | NP_001367493.1 | 3 - 4 | 252 | 85 | |
NM_068465.6 | NP_500866.2 | 4 - 5 | 430 | 85 | ||
NR_131524.1 | 3 - 4 | 252 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |