Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lntl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380241.1 | NP_001367493.1 | 5 - 6 | 633 | 116 | |
NM_001380242.1 | NP_001368410.1 | 2 - 3 | 171 | 116 | ||
NM_068465.6 | NP_500866.2 | 6 - 7 | 811 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |