Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373016.2 | NP_001360139.1 | 1 - 2 | 146 | 59 | |
NM_071630.5 | NP_504031.1 | 1 - 2 | 139 | 59 | ||
NM_071631.7 | NP_504032.1 | 1 - 2 | 146 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |