Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072750.6 | NP_505151.2 | 1 - 2 | 54 | 77 | |
NM_182343.6 | NP_872143.2 | 2 - 3 | 99 | 77 |
Gene Symbol: C08D8.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380704.1 | NP_001368232.1 | 1 - 2 | 60 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |