Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060583.4 | NP_492984.1 | 9 - 10 | 1620 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |