Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060630.6 | NP_493031.1 | 7 - 8 | 1614 | 86 | |
GenBank mRNA | AY559747.1 | - | 6 - 7 | 1464 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |