Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060656.4 | NP_493057.1 | 20 - 21 | 4937 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |