Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060690.1 | NP_493091.1 | 8 - 9 | 1785 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |