Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060733.2 | NP_493134.2 | 6 - 7 | 799 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |