Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060734.4 | NP_493135.2 | 5 - 6 | 684 | 86 | |
GenBank mRNA | AY037795.1 | - | 5 - 6 | 684 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |