Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047222.6 | NP_001040687.1 | 2 - 3 | 207 | 63 | |
GenBank mRNA | AF070968.1 | - | 2 - 3 | 206 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |