Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330940.2 | NP_001317729.1 | 2 - 3 | 318 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |