Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lagr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061002.3 | NP_493403.1 | 5 - 6 | 660 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |