Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zoo-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265447.3 | NP_001252376.1 | 1 - 2 | 102 | 66 | |
NM_001377679.2 | NP_001364648.1 | 1 - 2 | 104 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |