Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eva-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322695.3 | NP_001309519.1 | 8 | 983 | 71 | |
NM_001381268.1 | NP_001366909.1 | 7 - 8 | 1711 | 71 | ||
NM_061189.6 | NP_493590.2 | 7 - 8 | 974 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |