Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tre-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061248.5 | NP_493649.2 | 3 - 4 | 1770 | 61 | |
GenBank mRNA | AJ512339.1 | - | 3 - 4 | 1792 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |