Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prp-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061345.10 | NP_493746.1 | 1 - 2 | 107 | 122 | |
GenBank mRNA | U09415.1 | - | 1 - 2 | 118 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |