Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267093.2 | NP_001254022.1 | 5 - 6 | 743 | 67 | |
NM_001267094.2 | NP_001254023.1 | 2 - 3 | 383 | 67 | ||
NM_001318279.3 | NP_001305208.1 | 1 - 2 | 167 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |