Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxc-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129156.2 | NP_001122628.1 | 4 - 5 | 2490 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |