Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061496.2 | NP_493897.1 | 1 - 2 | 93 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |