Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jip-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038242.6 | NP_001033331.1 | 1 - 2 | 165 | 119 | |
NM_001377691.1 | NP_001364697.1 | 9 - 10 | 1593 | 119 | ||
NM_001377692.2 | NP_001364548.1 | 9 - 10 | 1599 | 119 | ||
NM_001377693.2 | NP_001364549.1 | 1 - 2 | 165 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |