Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047307.3 | NP_001040772.1 | 8 - 9 | 1667 | 88 | |
NM_001047308.1 | NP_001040773.1 | 8 - 9 | 1667 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |