Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061693.5 | NP_494094.1 | 1 - 2 | 85 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |