Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: math-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061709.1 | NP_494110.1 | 3 - 4 | 486 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |