Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381350.1 | NP_001368720.1 | 8 - 9 | 1577 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |