Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061971.5 | NP_494372.2 | 6 - 7 | 1044 | 91 | |
GenBank mRNA | AJ505020.1 | - | 6 - 7 | 985 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |