Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001361853.3 | NP_001348700.1 | 7 - 8 | 960 | 73 | |
NM_001361854.3 | NP_001348702.1 | 6 | 905 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |