Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcmo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062293.6 | NP_494694.2 | 4 - 5 | 1210 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |