Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lat-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047259.5 | NP_001040724.1 | 8 - 9 | 1232 | 77 | |
GenBank mRNA | AY314772.1 | - | 8 - 9 | 1215 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |