Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047261.5 | NP_001040726.1 | 1 - 2 | 115 | 78 | |
NM_062371.5 | NP_494772.1 | 4 - 5 | 431 | 78 | ||
NM_062372.6 | NP_494773.1 | 2 - 3 | 177 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |