Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062422.8 | NP_494823.2 | 1 - 2 | 135 | 127 | |
GenBank mRNA | AJ745074.1 | - | 1 - 2 | 133 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |