Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062545.8 | NP_494946.2 | 1 - 2 | 136 | 140 | |
NM_062546.8 | NP_494947.2 | 1 - 2 | 125 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |