Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062552.5 | NP_494953.3 | 13 - 14 | 1702 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |