Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062605.9 | NP_495006.2 | 10 - 11 | 1505 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |