Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062610.9 | NP_495011.1 | 2 - 3 | 324 | 102 | |
GenBank mRNA | AB007320.1 | - | 2 - 3 | 343 | 102 | |
AF025666.1 | - | 2 - 3 | 326 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |