Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062614.5 | NP_495015.1 | 3 - 4 | 831 | 111 |
Gene Symbol: rsp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381418.1 | NP_001367729.1 | 3 - 4 | 611 | 111 | |
NM_062613.3 | NP_495014.1 | 3 - 4 | 598 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |