Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026802.3 | NP_001021973.1 | 3 - 4 | 224 | 88 | |
NM_001381435.2 | NP_001367527.1 | 2 - 3 | 320 | 88 | ||
GenBank mRNA | AB112928.1 | - | 1 - 2 | 77 | 88 | |
U68265.1 | - | 1 - 2 | 99 | 88 | ||
U68266.1 | - | 2 - 3 | 161 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |