Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062741.7 | NP_495142.1 | 1 - 2 | 134 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |