Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062827.5 | NP_495228.1 | 1 - 2 | 598 | 103 | |
GenBank mRNA | AY887905.1 | - | 1 - 2 | 620 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |