Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ift-74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356863.3 | NP_001343748.1 | 5 - 6 | 1291 | 146 | |
NM_062958.6 | NP_495359.2 | 6 - 7 | 1536 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |