Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026958.5 | NP_001022129.1 | 7 - 8 | 1229 | 82 | |
NM_062993.8 | NP_495394.3 | 7 - 8 | 1230 | 82 | ||
GenBank mRNA | L15302.1 | - | 7 - 8 | 1251 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |