Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063124.7 | NP_495525.2 | 4 - 5 | 344 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |